missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-D-pipecolinic acid, 98%

Product Code. 15478417
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15478417 5 g 5g
15488417 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15478417 Supplier Thermo Scientific Alfa Aesar Supplier No. H51961.06
 Restricted Item
Estimated Shipment: 04-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 28697-17-8 | C11H19NO4 | 229.276 g/mol

It is also applied as catalytic agent and petrochemical additive. It is also an important organic intermediate used in agrochemicals, pharmaceuticals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is also applied as catalytic agent and petrochemical additive. It is also an important organic intermediate used in agrochemicals, pharmaceuticals and dyestuff fields.

Solubility
Soluble in 2ml dimethyl formamide 0.3gram.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 28697-17-8
Molecular Formula C11H19NO4
Molecular Weight (g/mol) 229.276
MDL Number MFCD00237380
InChI Key JQAOHGMPAAWWQO-MRVPVSSYSA-N
Synonym r-+-n-boc-2-piperidinecarboxylic acid, boc-d-hopro-oh, boc-d-pipecolic acid, r-1-tert-butoxycarbonyl piperidine-2-carboxylic acid, boc-d-pip-oh, d-n-boc-pipecolic acid, r-1-n-boc-pipecolinic acid, r-n-boc-piperidine-2-carboxylic acid, +-n-boc-r-pipecolinic acid, r-n-boc-2-piperidinecarboxylic acid
PubChem CID 688618
IUPAC Name (2R)-1-[(2-methylpropan-2-yl)oxycarbonyl]piperidine-2-carboxylic acid
SMILES CC(C)(C)OC(=O)N1CCCCC1C(=O)O

Specifications

Melting Point 116°C to 119°C
Quantity 5 g
Beilstein 480023
Solubility Information Soluble in 2ml dimethyl formamide 0.3gram.
Optical Rotation +68°, (c=1 in Acetic Acid)
Formula Weight 229.28
Percent Purity 98%
Chemical Name or Material N-Boc-D-pipecolinic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.