missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-D-phenylglycine, 99%

Product Code. 11339395
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11339395 1 g 1g
11349395 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11339395 Supplier Thermo Scientific Alfa Aesar Supplier No. L18540.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 33125-05-2 | C13H17NO4 | 251.28 g/mol

It is a reagent of choice for assignment of absolute configuration of chiral primary amines by 1H NMR, giving better results than Mosher's acid ((R)-(+)-Methoxy-(trifluoromethyl)­phenyl­acetic acid. αR)-α-[[(1,1-Dimethylethoxy)carbonyl]amino]-benzeneacetic Acid is D-(+)-2-Phenylglycine with Boc protecting group. (αR)-α-[[(1,1-Dimethylethoxy)carbonyl]amino]-benzeneacetic Acid is used as part of a catalyst combination to catalyze regioselective [4 + 2] cycloadditions of β-substituted cyclic enones and polyconjugated malononitriles. It can also be used to catalyze stereoselective preparation of polyfunctional nitrocyclohexene carboxaldehydes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a reagent of choice for assignment of absolute configuration of chiral primary amines by 1H NMR, giving better results than Mosher′s acid ((R)-(+)-ɑ-Methoxy-ɑ-(trifluoromethyl)­phenyl­acetic acid. αR)-α-[[(1,1-Dimethylethoxy)carbonyl]amino]-benzeneacetic Acid is D-(+)-2-Phenylglycine with Boc protecting group. (αR)-α-[[(1,1-Dimethylethoxy)carbonyl]amino]-benzeneacetic Acid is used as part of a catalyst combination to catalyze regioselective [4 + 2] cycloadditions of β-substituted cyclic enones and polyconjugated malononitriles. It can also be used to catalyze stereoselective preparation of polyfunctional nitrocyclohexene carboxaldehydes.

Solubility
Insoluble in water. Slightly soluble in DMSO and methanol.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 33125-05-2
Molecular Formula C13H17NO4
Molecular Weight (g/mol) 251.28
MDL Number MFCD00062043,MFCD00065588
InChI Key HOBFSNNENNQQIU-UHFFFAOYNA-N
Synonym boc-d-phg-oh, boc-d-phenylglycine, n-boc-d-phenylglycine, n-boc-d-2-phenylglycine, n-boc-l-phenylglycine, n-tert-butoxycarbonyl-d-2-phenylglycine, boc-d-alpha-phenylglycine, r-tert-butoxycarbonyl amino phenyl acetic acid, r-tert-butoxycarbonylamino-phenyl-acetic acid, r-2-tert-butoxycarbonyl amino-2-phenylacetic acid
PubChem CID 2755953
IUPAC Name (2R)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-2-phenylacetic acid
SMILES CC(C)(C)OC(=O)NC(C(O)=O)C1=CC=CC=C1

Specifications

Melting Point 90°C to 92°C
Beilstein 3033982
Solubility Information Insoluble in water. Slightly soluble in DMSO and methanol.
Quantity 1 g
Formula Weight 251.28
Optical Rotation −144° (c=1 in Ethanol)
Percent Purity 99%
Chemical Name or Material N-Boc-D-phenylglycine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.