missing translation for 'onlineSavingsMsg'
Learn More

N-BOC-D-Phenylalaninal, 97%, Thermo Scientific™

Product Code. 10435275 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10435275 1 g 1g
10678794 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10435275 Supplier Thermo Scientific Acros Supplier No. 376730010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 77119-85-8
Molecular Formula C14H19NO3
Molecular Weight (g/mol) 249.31
MDL Number MFCD00274187
InChI Key ZJTYRNPLVNMVPQ-GFCCVEGCSA-N
Synonym n-boc-d-phenylalaninal, boc-d-phenylalaninal, r-+-2-tert-butoxycarbonylamino-3-phenylpropanal, r-tert-butyl 1-oxo-3-phenylpropan-2-yl carbamate, n-boc-2 r-3-phenylpropanal, tert-butyl n-2r-1-oxo-3-phenylpropan-2-yl carbamate, tert-butyl 2r-1-oxo-3-phenylpropan-2-yl carbamate, carbamic acid, 1r-1-formyl-2-phenylethyl-, 1,1-dimethylethyl ester, boc-d-phe-h, pubchem11217
PubChem CID 7020444
IUPAC Name tert-butyl N-[(2R)-1-oxo-3-phenylpropan-2-yl]carbamate
SMILES CC(C)(C)OC(=O)N[C@H](CC1=CC=CC=C1)C=O

Specifications

Melting Point 85.0°C to 89.0°C
CAS Min % 96.0
CAS Max % 100.0
Color Tan or White
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass Bottle
Specific Rotation + 45.00
Specific Rotation Condition + 45.00 (20.00°C C=0.5,Methanol)
Quantity 1 g
Formula Weight 249.31
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material N-BOC-D-Phenylalaninal
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.