missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-D-glutamic acid 5-tert-butyl ester, 98%, Thermo Scientific Chemicals

Product Code. 15469718
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15469718 5 g 5g
15459718 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15469718 Supplier Thermo Scientific Alfa Aesar Supplier No. H63172.06
 Restricted Item
Estimated Shipment: 24-08-2026
Add to basket
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 104719-63-3
Molecular Formula C14H25NO6
Molecular Weight (g/mol) 303.355
MDL Number MFCD00076927
InChI Key YGSRAYJBEREVRB-SECBINFHSA-N
Synonym boc-d-glu otbu-oh, r-5-tert-butoxy-2-tert-butoxycarbonyl amino-5-oxopentanoic acid, boc-d-glutamic acid 5-tert-butyl ester, 2r-5-tert-butoxy-2-tert-butoxycarbonyl amino-5-oxopentanoic acid, boc-d-glu otbu-oh tlc, d-glutamic acid,n-1,1-dimethylethoxy carbonyl-,5-1,1-dimethylethyl ester
PubChem CID 7018748
IUPAC Name (2R)-5-[(2-methylpropan-2-yl)oxy]-2-[(2-methylpropan-2-yl)oxycarbonylamino]-5-oxopentanoic acid
SMILES CC(C)(C)OC(=O)CCC(C(=O)O)NC(=O)OC(C)(C)C

Specifications

Beilstein 3594191
Quantity 5 g
Formula Weight 303.35
Percent Purity 98%
Physical Form Solid
Chemical Name or Material N-Boc-D-glutamic acid 5-tert-butyl ester

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.