missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-D-glutamic acid 1-benzyl ester, 99%

Product Code. 11435047
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11435047

Brand: Thermo Scientific Alfa Aesar B21944.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Currently Unavailable: We have no information when this product will be available.
View alternative products
This item is not returnable. View return policy

CAS: 34404-30-3 | C17H23NO6 | 337.372 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 34404-30-3
Molecular Formula C17H23NO6
Molecular Weight (g/mol) 337.372
MDL Number MFCD00038266
InChI Key CVZUKWBYQQYBTF-CYBMUJFWSA-N
Synonym boc-d-glu-obzl, boc-d-glutamic acid 1-benzyl ester, n-boc-d-glutamic acid 1-benzyl ester, n-boc-d-glutamic acid benzyl ester, 1-benzyl n-boc-l-glutamate, n-tert-butoxycarbonyl-d-glutamic acid 1-benzyl ester, 4r-5-benzyloxy-4-tert-butoxycarbonyl amino-5-oxopentanoic acid, boc-d-glu-obn, pubchem12942, 1-benzyl n-boc-d-glutamate
PubChem CID 7010515
IUPAC Name (4R)-4-[(2-methylpropan-2-yl)oxycarbonylamino]-5-oxo-5-phenylmethoxypentanoic acid
SMILES CC(C)(C)OC(=O)NC(CCC(=O)O)C(=O)OCC1=CC=CC=C1

Specifications

Quantity 1 g
Formula Weight 337.37
Percent Purity 99%
Chemical Name or Material N-Boc-D-glutamic acid 1-benzyl ester

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.