missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-D-cyclohexylglycine, TRC

Product Code. 30342377
Change view
Click to view available options
Quantity:
2.5 g
Unit Size:
2.5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30342377 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30342377 Supplier Toronto Research Chemicals Supplier No. TRCB6462102.5G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-Boc-D-cyclohexylglycine
CAS 70491-05-3
Molecular Formula C13H23NO4
Quantity 2.5 g
Synonym (alphaR)-alpha-[[(1,1-Dimethylethoxy)carbonyl]amino]cyclohexaneacetic Acid, (2R)-2-[(tert-Butoxycarbonyl)amino]-2-(cyclohexyl)ethanoic Acid, (R)-2-(tert-Butoxycarbonylamino)-2-(cyclohexyl)acetic Acid, N-tert-Butoxycarbonyl-D-cyclohexylglycine
InChI Formula InChI=1S/C13H23NO4/c1-13(2,3)18-12(17)14-10(11(15)16)9-7-5-4-6-8-9/h9-10H,4-8H2,1-3H3,(H,14,17)(H,15,16)/t10-/m1/s1
SMILES CC(C)(C)OC(=O)N[C@H](C1CCCCC1)C(=O)O
IUPAC Name (2R)-2-cyclohexyl-2-[(2-methylpropan-2-yl)oxycarbonylamino]acetic acid
Molecular Weight (g/mol) 257.33
Formula Weight 257.1627
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.