missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-D-beta-prolinol, 97+%, Thermo Scientific™

Product Code. 15493738
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15493738 1 g 1g
15403748 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15493738 Supplier Thermo Scientific Alfa Aesar Supplier No. H57217.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

Solubility
Slightly soluble in water

Notes
Store in a cool ventilated place. Avoid strong oxidizing materials.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 138108-72-2
Molecular Formula C10H19NO3
Molecular Weight (g/mol) 201.266
MDL Number MFCD03094725
InChI Key HKIGXXRMJFUUKV-MRVPVSSYSA-N
Synonym r-tert-butyl 3-hydroxymethyl pyrrolidine-1-carboxylate, r-1-boc-3-hydroxymethyl pyrrolidine, r-1-boc-3-hydroxymethylpyrrolidine, tert-butyl 3r-3-hydroxymethyl pyrrolidine-1-carboxylate, r-3-hydroxymethyl-pyrrolidine-1-carboxylic acid tert-butyl ester, r-1-boc-3-hydroxymethyl-pyrrolidine, n-boc-d-beta-prolinol, r-boc-3-pyrrolidinemethanol, r-n-boc-pyrrolidine-3-methanol, r-3-hydroxymethylpyrrolidine-1-carboxylic acid tert-butyl ester
PubChem CID 1514340
IUPAC Name tert-butyl (3R)-3-(hydroxymethyl)pyrrolidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCC(C1)CO

Specifications

Density 1.089
Quantity 1 g
Solubility Information Slightly soluble in water
Formula Weight 201.27
Percent Purity ≥97%
Chemical Name or Material N-Boc-D-beta-prolinol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.