missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-D-beta-proline, 95%, Thermo Scientific Chemicals

Product Code. 15434238
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15434238 1 g 1g
15444238 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15434238 Supplier Thermo Scientific Alfa Aesar Supplier No. H57825.03
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

Solubility
Soluble in Dimethyl sulfoxide (DMSO).

Notes
Store in a cool ventilated place. Avoid strong oxidizing materials. Hygroscopic.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 72925-16-7
Molecular Formula C10H17NO4
Molecular Weight (g/mol) 215.25
MDL Number MFCD03094727
InChI Key HRMRQBJUFWFQLX-UHFFFAOYNA-N
Synonym r-1-n-boc-beta-proline, r-1-tert-butoxycarbonyl pyrrolidine-3-carboxylic acid, r-1-boc-pyrrolidine-3-carboxylic acid, r-1-boc-3-carboxyl pyrrolidine, boc-l-beta-proline, n-boc-d-beta-proline, r-n-boc-pyrrolidine-3-carboxylic acid, r-1-n-boc-pyrrolidine-3-carboxylic acid, 3r-1-tert-butoxycarbonyl pyrrolidine-3-carboxylic acid, 1,3-pyrrolidinedicarboxylic acid, 1-1,1-dimethylethyl ester, 3r
PubChem CID 1512491
SMILES CC(C)(C)OC(=O)N1CCC(C1)C(O)=O

Specifications

Quantity 1 g
Solubility Information Soluble in Dimethyl sulfoxide (DMSO).
Optical Rotation −15° (c=0.8 in Chloroform)
IUPAC Name (3R)-1-[(2-methylpropan-2-yl)oxycarbonyl]pyrrolidine-3-carboxylic acid
Formula Weight 215.25
Percent Purity 95%
Chemical Name or Material N-Boc-D-beta-proline

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.