missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-D-aspartic acid, 98%, Thermo Scientific™

Product Code. 15469288
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 15469288

Brand: Thermo Scientific Alfa Aesar H62820.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 62396-48-9
Molecular Formula C9H15NO6
Molecular Weight (g/mol) 233.22
MDL Number MFCD00798618
InChI Key KAJBMCZQVSQJDE-RXMQYKEDSA-N
Synonym boc-d-aspartic acid, boc-d-asp-oh, n-boc-d-aspartic acid, n-tert-butoxycarbonyl-d-aspartic acid, d-aspartic acid, n-1,1-dimethylethoxy carbonyl, 2r-2-tert-butoxycarbonyl amino butanedioic acid, boc-d-asparticacid, pubchem12159, n-t-boc-d-aspartic acid, ksc494c3p
PubChem CID 7021110
IUPAC Name (2R)-2-[(2-methylpropan-2-yl)oxycarbonylamino]butanedioic acid
SMILES CC(C)(C)OC(=O)NC(CC(=O)O)C(=O)O

Specifications

Quantity 25 g
Formula Weight 233.22
Percent Purity 98%
Physical Form Solid
Chemical Name or Material N-Boc-D-aspartic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.