missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-D-aspartic acid 4-benzyl ester, 98%

Product Code. 15448598
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 15448598

Brand: Thermo Scientific Alfa Aesar H62000.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 51186-58-4 | C16H21NO6 | 323.345 g/mol

N-Boc-D-aspartic acid 4-benzyl ester is used as an intermediate in organic synthesis and pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Boc-D-aspartic acid 4-benzyl ester is used as an intermediate in organic synthesis and pharmaceuticals.

Notes
Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51186-58-4
Molecular Formula C16H21NO6
Molecular Weight (g/mol) 323.345
MDL Number MFCD00038255
InChI Key SOHLZANWVLCPHK-GFCCVEGCSA-N
Synonym boc-d-asp obzl-oh, boc-d-aspartic acid 4-benzyl ester, r-4-benzyloxy-2-tert-butoxycarbonyl amino-4-oxobutanoic acid, 2r-4-benzyloxy-2-tert-butoxycarbonyl amino-4-oxobutanoic acid, n-alpha-t-boc-d-aspartic acid beta-benzyl ester, boc-d-asparticacid4-benzylester, pubchem11179, boc-d-asp obn-oh, boc-d-aspartic acid b-benzyl ester, boc-d-aspartic acid ?-benzyl ester
PubChem CID 6994663
IUPAC Name (2R)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-4-oxo-4-phenylmethoxybutanoic acid
SMILES CC(C)(C)OC(=O)NC(CC(=O)OCC1=CC=CC=C1)C(=O)O

Specifications

Quantity 5 g
Beilstein 2305471
Formula Weight 323.34
Percent Purity 98%
Chemical Name or Material N-Boc-D-aspartic acid 4-benzyl ester

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.