missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-D-alpha-phenylglycinol, 99%

Product Code. 11456300
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11456300 1 g 1g
11466300 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11456300 Supplier Thermo Scientific Alfa Aesar Supplier No. H27163.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 102089-74-7 | C13H19NO3 | 237.299 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 102089-74-7
Molecular Formula C13H19NO3
Molecular Weight (g/mol) 237.299
MDL Number MFCD00274205
InChI Key IBDIOGYTZBKRGI-NSHDSACASA-N
Synonym boc-d-phenylglycinol, r-n-tert-butoxycarbonyl-2-phenylglycinol, --n-boc-d-alpha-phenylglycinol, r-tert-butyl 2-hydroxy-1-phenylethyl carbamate, n-boc-d-2-phenylglycinol, r-2-tert-butoxycarbonylamino-2-phenylethanol, tert-butyl 1r-2-hydroxy-1-phenylethyl carbamate, tert-butyl n-1r-2-hydroxy-1-phenylethyl carbamate, carbamic acid, 1r-2-hydroxy-1-phenylethyl-, 1,1-dimethylethyl ester, r---2-boc-amino-2-phenylethanol
PubChem CID 7016461
IUPAC Name tert-butyl N-[(1R)-2-hydroxy-1-phenylethyl]carbamate
SMILES CC(C)(C)OC(=O)NC(CO)C1=CC=CC=C1

Specifications

Melting Point 137°C to 141°C
Quantity 1 g
Formula Weight 237.3
Optical Rotation −40° (c=1 in Chloroform)
Percent Purity 99%
Chemical Name or Material N-Boc-D-alpha-phenylglycinol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.