missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Boc-D-alaninol, TRC
Click to view available options
Quantity:
2.5 g
Unit Size:
2.5g
Specifications
Specifications
| Chemical Name or Material | N-Boc-D-alaninol |
| CAS | 106391-86-0 |
| Molecular Formula | C8H17NO3 |
| Quantity | 2.5 g |
| Synonym | Carbamic acid, N-[(1R)-2-hydroxy-1-methylethyl]-, 1,1-dimethylethyl ester (ACI), 1,1-Dimethylethyl N-[(1R)-2-hydroxy-1-methylethyl]carbamate (ACI), Carbamic acid, (2-hydroxy-1-methylethyl)-, 1,1-dimethylethyl ester, (R)- (ZCI), Carbamic acid, [(1R)-2-hydroxy-1-methylethyl]-, 1,1-dimethylethyl ester (9CI), ((1R)-2-Hydroxy-1-methylethyl)carbamic acid tert-butyl ester, ((R)-2-Hydroxy-1-methyl-ethyl)-carbamic acid tert-butyl ester, ((R)-2-Hydroxy-1-methylethyl)carbamic acid tert-butyl ester, (R)-(+)-2-(tert-Butoxycarbonylamino)-1-propanol, (R)-2-(N-tert-Butoxycarbonyl)aminopropan-1-ol, (R)-2-(tert-Butoxycarbonylamino)-1-propanol, (R)-2-(tert-Butoxycarbonylamino)-1-propanol, (R)-2-[(tert-Butyloxycarbonyl)amino]propan-1-ol, (R)-N-Boc-2-aminopropan-1-ol, 1,1-Dimethylethyl ((1R)-2-hydroxy-1-methylethyl)carbamate, Boc-D-alaninol, N-((R)-2-Hydroxy-1-methylethyl)carbamic acid tert-butyl ester, N-(tert-Butoxycarbonyl)-D-alaninol, tert-Butyl ((1R)-2-hydroxy-1-methylethyl)carbamate, tert-Butyl ((2R)-1-hydroxypropan-2-yl)carbamate, tert-Butyl ((R)-1-hydroxypropan-2-yl)carbamate, tert-Butyl (R)-(1-hydroxypropan-2-yl)carbamate, tert-Butyl N-((R)-2-hydroxy-1-methylethyl)carbamate, tert-Butyl N-[(2R)-1-hydroxypropan-2-yl]carbamate, N-tert-Butyloxycarbonyl D-Alaninol, N-Boc-D-alaninol |
| InChI Formula | InChI=1S/C8H17NO3/c1-6(5-10)9-7(11)12-8(2,3)4/h6,10H,5H2,1-4H3,(H,9,11)/t6-/m1/s1 |
| SMILES | C[C@H](CO)NC(=O)OC(C)(C)C |
| IUPAC Name | tert-butyl N-[(2R)-1-hydroxypropan-2-yl]carbamate |
| Molecular Weight (g/mol) | 175.23 |
| Formula Weight | 175.1208 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction