missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-D-alanine methyl ester, 98%, Thermo Scientific™

Product Code. 15418308
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 91103-47-8
Molecular Formula C9H17NO4
Molecular Weight (g/mol) 203.238
MDL Number MFCD00191865
InChI Key GJDICGOCZGRDFM-ZCFIWIBFSA-N
Synonym boc-d-ala-ome, boc-d-alanine methyl ester, r-methyl 2-tert-butoxycarbonyl amino propanoate, n-boc-d-alanine methyl ester, n-tert-butoxycarbonyl-d-alanine methyl ester, d-alanine, n-1,1-dimethylethoxy carbonyl-, methyl ester, methyl 2r-2-tert-butoxycarbonyl amino propanoate, boc-d-ala.ome, n-t-boc-d-alanine methyl ester
PubChem CID 637612
IUPAC Name methyl (2R)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoate
SMILES CC(C(=O)OC)NC(=O)OC(C)(C)C

Specifications

Melting Point 34°C to 37°C
Color White
Beilstein 4310313
Formula Weight 203.24
Percent Purity 98%
Density 1.030 g/mL
Quantity 1 g
Flash Point 113°C (235°F)
Physical Form Powder
Chemical Name or Material N-Boc-D-alanine methyl ester
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'