missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-cis-4-hydroxy-L-proline methyl ester, 97%, Thermo Scientific™

Product Code. 11491670
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
This item is not returnable. View return policy

Product Code. 11491670

Brand: Thermo Scientific Alfa Aesar H27018.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Used as a local anaesthetic. Used as therapeutic agents. Employed as important intermediates in various areas such as peptide synthesis, polymer chemistry..

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 102195-79-9
Molecular Formula C11H19NO5
Molecular Weight (g/mol) 245.275
MDL Number MFCD00237541
InChI Key MZMNEDXVUJLQAF-YUMQZZPRSA-N
Synonym n-boc-cis-4-hydroxy-l-proline methyl ester, boc-cis-hyp-ome, 2s,4s-1-tert-butyl 2-methyl 4-hydroxypyrrolidine-1,2-dicarboxylate, cis-boc-hyp-ome, boc-cis-4-hydroxy-l-proline methyl ester, 1-tert-butyl 2-methyl 2s,4s-4-hydroxypyrrolidine-1,2-dicarboxylate, methyl cis-1-boc-4-hydroxy-l-prolinate, 2s,4s-4-hydroxy-pyrrolidine-1,2-dicarboxylic acid 1-tert-butyl ester 2-methyl ester, methyl 4s-n-boc-4-hydroxy-l-prolinate, o1-tert-butyl o2-methyl 2s,4s-4-hydroxypyrrolidine-1,2-dicarboxylate
PubChem CID 10752873
IUPAC Name 1-O-tert-butyl 2-O-methyl (2S,4S)-4-hydroxypyrrolidine-1,2-dicarboxylate
SMILES CC(C)(C)OC(=O)N1CC(CC1C(=O)OC)O

Specifications

Melting Point 81°C to 84°C
Quantity 5 g
Solubility Information Insoluble in water.
Formula Weight 245.28
Percent Purity 97%
Chemical Name or Material N-Boc-cis-4-hydroxy-L-proline methyl ester
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.