missing translation for 'onlineSavingsMsg'
Learn More

N-BOC-cis-4-Hydroxy-L-proline, 97%

Product Code. 11936731 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11936731 5 g 5g
11926731 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11936731 Supplier Thermo Scientific Acros Supplier No. 441880050
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 87691-27-8 | C10H17NO5 | 231.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 87691-27-8
Molecular Formula C10H17NO5
Molecular Weight (g/mol) 231.25
MDL Number MFCD02094406
InChI Key BENKAPCDIOILGV-UHFFFAOYNA-N
Synonym n-boc-cis-4-hydroxy-l-proline, 2s,4s-1-tert-butoxycarbonyl-4-hydroxypyrrolidine-2-carboxylic acid, cis-1-tert-butoxycarbonyl-4-hydroxypyrrolidine-2-carboxylic acid, 2s,4s-cis-1-n-boc-4-hydroxy-proline, boc-cishyp-oh, boc-cis-4-hydroxy-l-proline, cis-n-boc-4-hydroxy-l-proline, unii-h4ycj0kw0u, h4ycj0kw0u, 4s-1-tert-butoxycarbonyl-4-hydroxy-l-proline
PubChem CID 688614
SMILES CC(C)(C)OC(=O)N1CC(O)CC1C(O)=O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 137.0°C to 143.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97%
Quantity 5 g
Specific Rotation − 49.60
IUPAC Name (2S,4S)-4-hydroxy-1-[(2-methylpropan-2-yl)oxycarbonyl]pyrrolidine-2-carboxylic acid
Formula Weight 231.25
Percent Purity 97%
Physical Form Crystalline Powder or Powder
Chemical Name or Material N-BOC-cis-4-Hydroxy-L-proline
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • N-BOC-cis-4-Hydroxy-L-proline
Signal Word
  • Warning
Hazard Category
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H400-Very toxic to aquatic life.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.