missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-cis-4-hydroxy-D-proline methyl ester, 95%

Product Code. 15489658
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15489658 1 g 1g
15428146 250 mg 250mg
15499658 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15489658 Supplier Thermo Scientific Alfa Aesar Supplier No. H63136.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 114676-69-6 | C11H19NO5 | 245.275 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 114676-69-6
Molecular Formula C11H19NO5
Molecular Weight (g/mol) 245.275
MDL Number MFCD00797548
InChI Key MZMNEDXVUJLQAF-HTQZYQBOSA-N
Synonym 2r,4r-1-tert-butyl 2-methyl 4-hydroxypyrrolidine-1,2-dicarboxylate, methyl cis-1-boc-4-hydroxy-d-prolinate, n-boc-cis-4-hydroxy-d-proline methyl ester, 1-tert-butyl 2-methyl 2r,4r-4-hydroxypyrrolidine-1,2-dicarboxylate, methyl 2r,4r-1-boc-4-hydroxypyrrolidine-2-carboxylate, 2r,4r-4-hydroxy-pyrrolidine-1,2-dicarboxylic acid 1-tert-butyl ester 2-methyl ester, 2r,4r-1-tert-butyl 2-methyl 4-hydroxypyrrolidine, o1-tert-butyl o2-methyl 2r,4r-4-hydroxypyrrolidine-1,2-dicarboxylate, pubchem14128, boc-cis-4-d-hyp-ome
PubChem CID 6951202
IUPAC Name 1-O-tert-butyl 2-O-methyl (2R,4R)-4-hydroxypyrrolidine-1,2-dicarboxylate
SMILES CC(C)(C)OC(=O)N1CC(CC1C(=O)OC)O

Specifications

Color White
Quantity 1 g
Formula Weight 245.27
Percent Purity 95%
Physical Form Powder
Chemical Name or Material N-Boc-cis-4-hydroxy-D-proline methyl ester

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.