missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Boc-γ-aminobutyric Acid, TRC
Specifications
Specifications
| Chemical Name or Material | N-Boc-Gamma-aminobutyric Acid |
| CAS | 57294-38-9 |
| Molecular Formula | C9H17NO4 |
| Quantity | 2.5 g |
| Synonym | Boc-4-aminobutanoic Acid, N-Boc-4-Aminobutyric Acid, N-Boc-γ-aminobutyric Acid N-tert-Butoxycarbonyl-γ-aminobutyric Acid, NSC 164040, 4-(tert-Butoxycarbonyl)aminobutyric Acid, 4-(tert-Butyloxycarbonylamino)butanoic Acid, 4-[(tert-Butoxycarbonyl)amino]butanoic Acid 4-[N-(tert-Butoxycarbonyl)amino]butyric Acid, 4-[[(1,1-Dimethylethoxy)carbonyl]amino]butanoic acid |
| InChI Formula | InChI=1S/C9H17NO4/c1-9(2,3)14-8(13)10-6-4-5-7(11)12/h4-6H2,1-3H3,(H,10,13)(H,11,12) |
| SMILES | CC(C)(OC(NCCCC(O)=O)=O)C |
| Molecular Weight (g/mol) | 203.24 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?