missing translation for 'onlineSavingsMsg'
Learn More

N-BOC-Amino-(4-hydroxycyclohexyl)carboxylic acid, 95%, Thermo Scientific™

Catalog Number p-3735666
Click to view available options
Quantity:
500 mg
Packaging:
Glass bottle
This item is not returnable. View return policy
369403-08-7
MFCD02683153
DRBVVUBYEBFRKS-UHFFFAOYSA-N
1-tert-butoxy carbonyl amino-4-hydroxycyclohexane-1-carboxylic acid, n-boc-amino-4-hydroxycyclohexyl carboxylic acid, 1-tert-butoxycarbonyl amino-4-hydroxycyclohexanecarboxylic acid, 1-tert-butoxycarbonyl amino-4-hydroxycyclohexane-1-carboxylic acid, 4-hydroxy-1-2-methylpropan-2-yl oxycarbonylamino cyclohexane-1-carboxylic acid, boc 4-cis/trans-1, 1-acch 4-oh, tertbutoxycarbonylaminohydroxycyclohexanecarboxylicacid, 1-boc-amino-4-hydroxy-cyclohexanecarboxylic acid
2734594
4-hydroxy-1-[(2-methylpropan-2-yl)oxycarbonylamino]cyclohexane-1-carboxylic acid
CC(C)(C)OC(=O)NC1(CCC(CC1)O)C(=O)O
This item is not returnable. View return policy
369403-08-7
MFCD02683153
DRBVVUBYEBFRKS-UHFFFAOYSA-N
1-tert-butoxy carbonyl amino-4-hydroxycyclohexane-1-carboxylic acid, n-boc-amino-4-hydroxycyclohexyl carboxylic acid, 1-tert-butoxycarbonyl amino-4-hydroxycyclohexanecarboxylic acid, 1-tert-butoxycarbonyl amino-4-hydroxycyclohexane-1-carboxylic acid, 4-hydroxy-1-2-methylpropan-2-yl oxycarbonylamino cyclohexane-1-carboxylic acid, boc 4-cis/trans-1, 1-acch 4-oh, tertbutoxycarbonylaminohydroxycyclohexanecarboxylicacid, 1-boc-amino-4-hydroxy-cyclohexanecarboxylic acid
2734594
4-hydroxy-1-[(2-methylpropan-2-yl)oxycarbonylamino]cyclohexane-1-carboxylic acid
CC(C)(C)OC(=O)NC1(CCC(CC1)O)C(=O)O

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
CAS Min % 94.0
CAS Max % 100.0
Color White
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass bottle
Molecular Formula C12H21NO5
Quantity 500 mg
Molecular Weight (g/mol) 259.3
Formula Weight 259.3
Percent Purity 95%
Physical Form Powder
Chemical Name or Material N-BOC-Amino-(4-hydroxycyclohexyl)carboxylic acid
Show More Show Less
compliance-icons
Product Identifier
  • N-BOC-Amino-(4-hydroxycyclohexyl)carboxylic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'