missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-5-oxo-L-proline, 97%

Product Code. p-7039581
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
This item is not returnable. View return policy

Product Code. 15405078

Brand: Thermo Scientific Alfa Aesar H58884.MD

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 53100-44-0 | C10H15NO5 | 229.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 53100-44-0
Molecular Formula C10H15NO5
Molecular Weight (g/mol) 229.23
MDL Number MFCD00672316
InChI Key MJLQPFJGZTYCMH-LURJTMIESA-N
Synonym boc-pyr-oh, n-boc-l-pyroglutamic acid, s-boc-5-oxopyrrolidine-2-carboxylic acid, boc-l-pyroglutamic acid, 2s-1-tert-butoxy carbonyl-5-oxopyrrolidine-2-carboxylic acid, s-1-tert-butoxycarbonyl-5-oxopyrrolidine-2-carboxylic acid, 1,2-pyrrolidinedicarboxylic acid, 5-oxo-, 1-1,1-dimethylethyl ester, 2s, 1-tert-butoxycarbonyl-5-oxo-l-proline, bocpyr, 2s-1-tert-butoxycarbonyl-5-oxopyrrolidine-2-carboxylic acid
PubChem CID 104412
IUPAC Name (2S)-1-[(2-methylpropan-2-yl)oxycarbonyl]-5-oxopyrrolidine-2-carboxylic acid
SMILES CC(C)(C)OC(=O)N1[C@@H](CCC1=O)C(O)=O

Specifications

Melting Point 118°C to 123°C
Quantity 250 mg
Sensitivity Moisture sensitive
Optical Rotation −35° (c=1 in Chloroform)
Formula Weight 229.23
Percent Purity 97%
Chemical Name or Material N-Boc-5-oxo-L-proline

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.