missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-5-oxo-D-proline, 97%

Product Code. 15494918
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
This item is not returnable. View return policy

Product Code. 15494918

Brand: Thermo Scientific Alfa Aesar H58701.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 160347-90-0 | C10H15NO5 | 229.232 g/mol

Used as pharmaceutical intermediates and anesthetic agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used as pharmaceutical intermediates and anesthetic agent.

Solubility
Slightly soluble in water.

Notes
Moisture Sensitive, store away from moisture. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 160347-90-0
Molecular Formula C10H15NO5
Molecular Weight (g/mol) 229.232
MDL Number MFCD08458561
InChI Key MJLQPFJGZTYCMH-ZCFIWIBFSA-N
Synonym boc-d-pyr-oh, r-boc-5-oxopyrrolidine-2-carboxylic acid, r-1-tert-butoxycarbonyl-5-oxopyrrolidine-2-carboxylic acid, n-boc-d-pyroglutamic acid, 2r-1-tert-butoxy carbonyl-5-oxopyrrolidine-2-carboxylic acid, 2r-1-tert-butoxycarbonyl-5-oxopyrrolidine-2-carboxylic acid, boc-d-pglu-oh, ambotzbaa5600, n-boc-5-oxo-d-proline
PubChem CID 18645552
IUPAC Name (2R)-1-[(2-methylpropan-2-yl)oxycarbonyl]-5-oxopyrrolidine-2-carboxylic acid
SMILES CC(C)(C)OC(=O)N1C(CCC1=O)C(=O)O

Specifications

Melting Point 111°C to 116°C
Solubility Information Slightly soluble in water.
Quantity 1 g
Formula Weight 229.23
Optical Rotation +35° (c=1 in Chloroform)
Sensitivity Moisture sensitive
Percent Purity 97%
Chemical Name or Material N-Boc-5-oxo-D-proline

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.