Learn More
N-BOC-4-Piperidone, 99%, Thermo Scientific Chemicals
£43.50 - £385.00
Chemical Identifiers
CAS | 79099-07-3 |
---|---|
Molecular Formula | C10H17NO3 |
Molecular Weight (g/mol) | 199.25 |
MDL Number | MFCD00151800 |
InChI Key | ROUYFJUVMYHXFJ-UHFFFAOYSA-N |
Synonym | 1-boc-4-piperidone, n-boc-4-piperidone, n-tert-butoxycarbonyl-4-piperidone, 1-boc-4-piperidinone, boc-4-piperidone, 1-tert-butoxycarbonyl-4-piperidone, n-boc-4-piperidinone, boc-piperidone, 1-t-boc-4-piperidone |
PubChem CID | 735900 |
IUPAC Name | tert-butyl 4-oxopiperidine-1-carboxylate |
SMILES | CC(C)(C)OC(=O)N1CCC(=O)CC1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10584911
|
Thermo Scientific Acros
330270050 |
5 g | Glass bottle |
£43.50
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10476432
|
Thermo Scientific Acros
330270250 |
25 g | Glass bottle |
£141.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11956321
|
Thermo Scientific Acros
330271000 |
100 g | Glass bottle |
£385.00
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
79099-07-3 | |
199.25 | |
ROUYFJUVMYHXFJ-UHFFFAOYSA-N | |
735900 | |
CC(C)(C)OC(=O)N1CCC(=O)CC1 |
C10H17NO3 | |
MFCD00151800 | |
1-boc-4-piperidone, n-boc-4-piperidone, n-tert-butoxycarbonyl-4-piperidone, 1-boc-4-piperidinone, boc-4-piperidone, 1-tert-butoxycarbonyl-4-piperidone, n-boc-4-piperidinone, boc-piperidone, 1-t-boc-4-piperidone | |
tert-butyl 4-oxopiperidine-1-carboxylate |
Specifications
79099-07-3 | |
100.0 | |
White to Yellow | |
98.5% min. (GC) | |
MFCD00151800 | |
Solubility in water: slightly soluble | |
CC(C)(C)OC(=O)N1CCC(=O)CC1 | |
199.25 | |
199.25 | |
Crystalline Powder |
98.5 | |
74.0°C to 75.0°C | |
Authentic | |
C10H17NO3 | |
1-boc-4-piperidone, n-boc-4-piperidone, n-tert-butoxycarbonyl-4-piperidone, 1-boc-4-piperidinone, boc-4-piperidone, 1-tert-butoxycarbonyl-4-piperidone, n-boc-4-piperidinone, boc-piperidone, 1-t-boc-4-piperidone | |
ROUYFJUVMYHXFJ-UHFFFAOYSA-N | |
tert-butyl 4-oxopiperidine-1-carboxylate | |
735900 | |
99% | |
N-BOC-4-Piperidone |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER
GHS Signal Word: Warning
- N-BOC-4-Piperidone
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H302-Harmful if swallowed.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only