missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-4-phenyl-L-phenylalanine, TRC

Product Code. 30342533
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30342533 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30342533 Supplier Toronto Research Chemicals Supplier No. TRCB6428001G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-Boc-4-phenyl-L-phenylalanine
CAS 147923-08-8
Molecular Formula C20H23NO4
Quantity 1 g
Synonym Boc-4-phenyl-Phe-OH, (S)-alpha-[[(1,1-Dimethylethoxy)carbonyl]amino]-[1,1'-Biphenyl]-4-propanoic Acid, (2S)-2-(tert-Butoxycarbonylamino)-3-(biphenyl-4-yl)propanoic Acid, (S)-2-tert-Butoxycarbonylamino-3-(biphenyl-4-yl)propionic Acid, N-tert-Butoxycarbonyl-(4-phenylphenyl)alanine, N-tert-Butoxycarbonyl-(S)-(4-biphenylyl)alanine
InChI Formula InChI=1S/C20H23NO4/c1-20(2,3)25-19(24)21-17(18(22)23)13-14-9-11-16(12-10-14)15-7-5-4-6-8-15/h4-12,17H,13H2,1-3H3,(H,21,24)(H,22,23)/t17-/m0/s1
SMILES CC(C)(C)OC(=O)N[C@@H](Cc1ccc(cc1)c2ccccc2)C(=O)O
IUPAC Name (2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-3-(4-phenylphenyl)propanoic acid
Molecular Weight (g/mol) 341.4
Formula Weight 341.1627
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.