missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-4-iodo-L-phenylalanine, 98%

Product Code. 15458417
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15458417 5 g 5g
15468417 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15458417 Supplier Thermo Scientific Alfa Aesar Supplier No. H51960.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 62129-44-6 | C14H18INO4 | 391.21 g/mol

N-Boc-4-iodo-L-phenylalanine is used as pharmaceutical intermediate. It is also used as a p-iodo phenyl alanine derivative with N-Boc protection.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Boc-4-iodo-L-phenylalanine is used as pharmaceutical intermediate. It is also used as a p-iodo phenyl alanine derivative with N-Boc protection.

Solubility
Slightly soluble in water.

Notes
Light sensitive. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 62129-44-6
Molecular Formula C14H18INO4
Molecular Weight (g/mol) 391.21
MDL Number MFCD00037119
InChI Key JZLZDBGQWRBTHN-NSHDSACASA-N
Synonym boc-4-iodo-l-phenylalanine, boc-phe 4-i-oh, boc-l-4-iodophenylalanine, boc-l-phe 4-i-oh, s-2-tert-butoxycarbonyl amino-3-4-iodophenyl propanoic acid, boc-4'-iodo-l-phe, 2s-2-tert-butoxy carbonyl amino-3-4-iodophenyl propanoic acid, 2s-2-tert-butoxycarbonyl amino-3-4-iodophenyl propanoic acid, l-n-tert-butoxycarbonyl-4-iodophenylalanine, n-tert-butoxycarbonyl-4-iodo-l-phenylalanine
PubChem CID 2755956
SMILES CC(C)(C)OC(=O)N[C@@H](CC1=CC=C(I)C=C1)C(O)=O

Specifications

Melting Point 150°C (decomposition)
Quantity 5 g
Beilstein 4503851
Sensitivity Light sensitive
Solubility Information Slightly soluble in water.
Optical Rotation +22.5° (c=1 in Ethyl Acetate)
IUPAC Name (2S)-3-(4-iodophenyl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
Formula Weight 391.21
Percent Purity 98%
Chemical Name or Material N-Boc-4-iodo-L-phenylalanine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.