missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-4-iodo-L-phenylalanine, 98%

Product Code. 15468417
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 15468417

Brand: Thermo Scientific Alfa Aesar H51960.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 62129-44-6 | C14H18INO4 | 391.21 g/mol

N-Boc-4-iodo-L-phenylalanine is used as pharmaceutical intermediate. It is also used as a p-iodo phenyl alanine derivative with N-Boc protection.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Boc-4-iodo-L-phenylalanine is used as pharmaceutical intermediate. It is also used as a p-iodo phenyl alanine derivative with N-Boc protection.

Solubility
Slightly soluble in water.

Notes
Light sensitive. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 62129-44-6
Molecular Formula C14H18INO4
Molecular Weight (g/mol) 391.21
MDL Number MFCD00037119
InChI Key JZLZDBGQWRBTHN-NSHDSACASA-N
Synonym boc-4-iodo-l-phenylalanine, boc-phe 4-i-oh, boc-l-4-iodophenylalanine, boc-l-phe 4-i-oh, s-2-tert-butoxycarbonyl amino-3-4-iodophenyl propanoic acid, boc-4'-iodo-l-phe, 2s-2-tert-butoxy carbonyl amino-3-4-iodophenyl propanoic acid, 2s-2-tert-butoxycarbonyl amino-3-4-iodophenyl propanoic acid, l-n-tert-butoxycarbonyl-4-iodophenylalanine, n-tert-butoxycarbonyl-4-iodo-l-phenylalanine
PubChem CID 2755956
SMILES CC(C)(C)OC(=O)N[C@@H](CC1=CC=C(I)C=C1)C(O)=O

Specifications

Melting Point 150°C (decomposition)
Quantity 25 g
Beilstein 4503851
Sensitivity Light sensitive
Solubility Information Slightly soluble in water.
Optical Rotation +22.5° (c=1 in Ethyl Acetate)
IUPAC Name (2S)-2-{[(tert-butoxy)carbonyl]amino}-3-(4-iodophenyl)propanoic acid
Formula Weight 391.21
Percent Purity 98%
Chemical Name or Material N-Boc-4-iodo-L-phenylalanine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.