missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-4-cyano-L-phenylalanine, 95%

Product Code. 15468618
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15468618 5 g 5g
15458618 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15468618 Supplier Thermo Scientific Alfa Aesar Supplier No. H62023.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 131724-45-3 | C15H18N2O4 | 290.319 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 131724-45-3
Molecular Formula C15H18N2O4
Molecular Weight (g/mol) 290.319
MDL Number MFCD00672526
InChI Key RMBLTLXJGNILPG-LBPRGKRZSA-N
Synonym boc-phe 4-cn-oh, boc-l-4-cyanophenylalanine, boc-4-cyano-l-phenylalanine, s-2-tert-butoxycarbonyl amino-3-4-cyanophenyl propanoic acid, boc-d-4-cyanophenylalanine, n-boc-4-cyanophenylalanine, s-n-boc-4-cyanophenylalanine, 2s-2-tert-butoxycarbonyl amino-3-4-cyanophenyl propanoic acid, 2s-3-4-cyanophenyl-2-2-methylpropan-2-yl oxycarbonylamino propanoic acid, boc-l-4-cn-phe-oh
PubChem CID 7020841
IUPAC Name (2S)-3-(4-cyanophenyl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
SMILES CC(C)(C)OC(=O)NC(CC1=CC=C(C=C1)C#N)C(=O)O

Specifications

Melting Point 156°C to 160°C
Color White
UN Number UN3439
Beilstein 7715793
Quantity 5 g
Formula Weight 290.31
Percent Purity 95%
Physical Form Powder
Chemical Name or Material N-Boc-4-cyano-L-phenylalanine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.