missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-3-nitro-L-phenylalanine, 95%, Thermo Scientific Chemicals

Catalog Number 15498557
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
131980-29-5
C14H18N2O6
310.306
MFCD01317033
OWTGPXDXLMNQKK-NSHDSACASA-N
boc-3-nitro-l-phenylalanine, boc-l-3-nitrophenylalanine, boc-phe 3-no2-oh, s-n-boc-3-nitrophenylalanine, 2s-2-tert-butoxycarbonyl amino-3-3-nitrophenyl propanoic acid, s-2-tert-butoxycarbonyl amino-3-3-nitrophenyl propanoic acid, boc-l-3-nitrophe, boc-l-3-no2-phe-oh, 2-tert-butoxycarbonylamino-3-3-nitro-phenyl-propionic acid
2761816
CC(C)(C)OC(=O)NC(CC1=CC(=CC=C1)[N+](=O)[O-])C(=O)O
This item is not returnable. View return policy
131980-29-5
C14H18N2O6
310.306
MFCD01317033
OWTGPXDXLMNQKK-NSHDSACASA-N
boc-3-nitro-l-phenylalanine, boc-l-3-nitrophenylalanine, boc-phe 3-no2-oh, s-n-boc-3-nitrophenylalanine, 2s-2-tert-butoxycarbonyl amino-3-3-nitrophenyl propanoic acid, s-2-tert-butoxycarbonyl amino-3-3-nitrophenyl propanoic acid, boc-l-3-nitrophe, boc-l-3-no2-phe-oh, 2-tert-butoxycarbonylamino-3-3-nitro-phenyl-propionic acid
2761816
CC(C)(C)OC(=O)NC(CC1=CC(=CC=C1)[N+](=O)[O-])C(=O)O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Formula Weight 310.31
Percent Purity 95%
Chemical Name or Material N-Boc-3-nitro-L-phenylalanine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'