missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-3-iodo-L-alanine methyl ester, 98%

Product Code. 15428198
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15428198 5 g 5g
15438198 250 mg 250mg
15418198 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15428198 Supplier Thermo Scientific Alfa Aesar Supplier No. H61638.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 93267-04-0 | C9H16INO4 | 329.13 g/mol

N-Boc-3-iodo-L-alanine methyl ester is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Boc-3-iodo-L-alanine methyl ester is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

Solubility
Soluble in water and 1% acetic acid.

Notes
Light sensitive. Store in dark. Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 93267-04-0
Molecular Formula C9H16INO4
Molecular Weight (g/mol) 329.13
MDL Number MFCD00216579
InChI Key UGZBFCCHLUWCQI-LURJTMIESA-N
Synonym boc-beta-iodo-ala-ome, boc-3-iodo-l-alanine methyl ester, r-methyl 2-tert-butoxycarbonyl amino-3-iodopropanoate, n-tert-butoxycarbonyl-3-iodo-l-alanine methyl ester, l-n-boc-3-iodoalanine methyl ester, r-methyl 2-tert-butoxycarbonylamino-3-iodopropanoate, boc-3-iodo-l-ala-ome, n-boc-3-iodo-l-alanine methyl ester, r-boc-, a-iodo-ala-ome, methyl 2r-2-tert-butoxycarbonyl amino-3-iodopropanoate
PubChem CID 10903591
SMILES COC(=O)[C@H](CI)NC(=O)OC(C)(C)C

Specifications

Melting Point 50°C to 52°C
Color White
Beilstein 4422011
Solubility Information Soluble in water and 1% acetic acid.
IUPAC Name methyl (2R)-3-iodo-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoate
Quantity 5 g
Formula Weight 329.13
Sensitivity Light sensitive
Percent Purity 98%
Physical Form Powder
Chemical Name or Material N-Boc-3-iodo-L-alanine methyl ester
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.