missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-3-chloro-L-phenylalanine, 95%

Product Code. 15488497
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. Quantity unitSize
15488497 5 g 5g
15478497 1 g 1g
2 options
This item is not returnable. View return policy

Product Code. 15488497

Brand: Thermo Scientific Alfa Aesar H52015.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 114873-03-9 | C14H18ClNO4 | 299.751 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 114873-03-9
Molecular Formula C14H18ClNO4
Molecular Weight (g/mol) 299.751
MDL Number MFCD00672515
InChI Key RCZHBTHQISEPPP-NSHDSACASA-N
Synonym boc-3-chloro-l-phenylalanine, boc-phe 3-cl-oh, boc-l-3-chlorophenylalanine, s-n-boc-3-chlorophenylalanine, boc-l-phe 3-cl-oh, boc-l-3-chlorophe, boc-d-3-chlorophenylalanine, boc-l-3-chloro-phe-oh, s-2-tert-butoxycarbonyl amino-3-3-chlorophenyl propanoic acid, n-boc-3-chloro-l-phenylalanine
PubChem CID 2761465
IUPAC Name (2S)-3-(3-chlorophenyl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
SMILES CC(C)(C)OC(=O)NC(CC1=CC(=CC=C1)Cl)C(=O)O

Specifications

Quantity 5 g
Formula Weight 299.75
Percent Purity 95%
Chemical Name or Material N-Boc-3-chloro-L-phenylalanine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.