missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-3-bromo-D-phenylalanine, 98%, Thermo Scientific™

Product Code. 15499238
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 261360-77-4
Molecular Formula C14H18BrNO4
Molecular Weight (g/mol) 344.205
MDL Number MFCD01317704
InChI Key FBUDYESOPLBQIR-LLVKDONJSA-N
Synonym boc-3-bromo-d-phenylalanine, r-n-boc-3-bromophenylalanine, boc-d-3-bromophenylalanine, r-3-3-bromophenyl-2-tert-butoxycarbonyl amino propanoic acid, boc-d-3-br-phe-oh, boc-d-3-bromophe, 2r-3-3-bromophenyl-2-tert-butoxycarbonyl amino propanoic acid, boc-d-phe 3-br-oh, r-boc-2-amino-3-3-bromophenyl propionic acid, d-phenylalanine, 3-bromo-n-1,1-dimethylethoxy carbonyl
PubChem CID 7006629
IUPAC Name (2R)-3-(3-bromophenyl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
SMILES CC(C)(C)OC(=O)NC(CC1=CC(=CC=C1)Br)C(=O)O

Specifications

Color White
Formula Weight 344.2
Percent Purity 98%
Quantity 250 mg
Physical Form Powder
Chemical Name or Material N-Boc-3-bromo-D-phenylalanine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'