missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-3-benzyloxymethyl-L-histidine, 98%, Thermo Scientific™

Product Code. 15487738
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79950-65-5
MDL Number MFCD00043133
Synonym Boc-His(Bom)-OH; N-[(1,1-Dimethylethoxy)carbonyl]-3-[(phenylmethoxy)methyl]- L-histidine

Specifications

Color White
Beilstein 4335455
Formula Weight 375.42
Percent Purity 98%
Quantity 5 g
Physical Form Powder
Chemical Name or Material N-Boc-3-benzyloxymethyl-L-histidine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'