missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-3-(1-naphthyl)-L-alanine, 96%, Thermo Scientific™

Product Code. 15448478
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 15448478

Brand: Thermo Scientific Alfa Aesar H61889.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 55447-00-2
Molecular Formula C18H21NO4
Molecular Weight (g/mol) 315.369
MDL Number MFCD00085377
InChI Key KHHIGWRTNILXLL-HNNXBMFYSA-N
Synonym boc-3-1-naphthyl-l-alanine, boc-l-1-naphthylalanine, boc-1-nal-oh, s-n-boc-l-1-naphthyl alanine, boc-l-3-1-naphthyl-alanine, boc-d-1-nal-oh, boc-3-1-naphthyl-l-ala, 2s-2-tert-butoxy carbonylamino-3-naphthylpropanoic acid, 3-naphthalen-1-yl-n-boc-l-alanine, boc-d-3-1-naphthyl-alanine
PubChem CID 7021841
IUPAC Name (2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-3-naphthalen-1-ylpropanoic acid
SMILES CC(C)(C)OC(=O)NC(CC1=CC=CC2=CC=CC=C21)C(=O)O

Specifications

Beilstein 3561332
Quantity 1 g
Formula Weight 315.36
Percent Purity 96%
Chemical Name or Material N-Boc-3-(1-naphthyl)-L-alanine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.