missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-2-pyrrolidinone, TRC

Product Code. 30346630
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30346630 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30346630 Supplier Toronto Research Chemicals Supplier No. TRCB66594510G
 Restricted Item
Estimated Shipment: 11-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-Boc-2-pyrrolidinone
CAS 85909-08-6
Molecular Formula C9H15NO3
Quantity 10 g
Synonym 2-Oxo-1-pyrrolidinecarboxylic Acid 1,1-Dimethylethyl Ester, 1,1-Dimethylethyl 2-oxo-1-pyrrolidinecarboxylate, 1-(tert-Butoxycarbonyl)pyrrolidin-2-one, 2-Oxopyrrolidine-1-carboxylic Acid tert-Butyl Ester, N-(tert-Butoxycarbonyl)pyrrolidin-2-one, N-(tert-Butyloxycarbonyl)pyrrolidin-2-one, N-Boc-2-pyrrolidinone, N-tert-Butoxycarbonyl-2-oxopyrrolidine, NSC 662770, tert-Butyl 2-Oxopyrrolidine-1-carboxylate
InChI Formula InChI=1S/C9H15NO3/c1-9(2,3)13-8(12)10-6-4-5-7(10)11/h4-6H2,1-3H3
SMILES CC(C)(C)OC(=O)N1CCCC1=O
IUPAC Name tert-butyl 2-oxopyrrolidine-1-carboxylate
Molecular Weight (g/mol) 185.22
Formula Weight 185.1052
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.