missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-2-cyclopropyl-L-glycine, 95%, Thermo Scientific Chemicals

Product Code. 15469358
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15469358 1 g 1g
15479358 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15469358 Supplier Thermo Scientific Alfa Aesar Supplier No. H62914.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 155976-13-9
Molecular Formula C10H17NO4
Molecular Weight (g/mol) 215.25
MDL Number MFCD06659132,MFCD01861823
InChI Key QFVJNEASAAJIDF-ZETCQYMHSA-N
Synonym boc-l-cyclopropylglycine, s-2-tert-butoxycarbonyl amino-2-cyclopropylacetic acid, boc-s-cyclopropylglycine, s-tert-butoxycarbonylamino-cyclopropyl-acetic acid, s-boc-amino-cyclopropylacetic acid, 2s-2-amino-2-cyclopropylethanoic acid, n-boc protected, s-2-tert-butoxycarbonylamino-2-cyclopropylacetic acid, s-tert-butoxycarbonyl amino cyclopropyl acetic acid, boc-l-cyclopropyl glycine
PubChem CID 2761461
IUPAC Name (2S)-2-cyclopropyl-2-[(2-methylpropan-2-yl)oxycarbonylamino]acetic acid
SMILES CC(C)(C)OC(=O)N[C@@H](C1CC1)C(O)=O

Specifications

Color White
Beilstein 6650020
Quantity 1 g
Formula Weight 215.25
Percent Purity 95%
Physical Form Powder
Chemical Name or Material N-Boc-2-cyclopropyl-L-glycine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.