missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-2-aminoisobutyric acid, 98+%

Product Code. p-7047589
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. Quantity unitSize
11398826 1 g 1g
11308836 5 g 5g
2 options
This item is not returnable. View return policy

Product Code. 11398826

Brand: Thermo Scientific Alfa Aesar L19484.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 30992-29-1 | C9H17NO4 | 203.238 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 30992-29-1
Molecular Formula C9H17NO4
Molecular Weight (g/mol) 203.238
MDL Number MFCD00042973
InChI Key MFNXWZGIFWJHMI-UHFFFAOYSA-N
Synonym boc-aib-oh, boc-alpha-methylalanine, 2-boc-amino isobutyric acid, 2-tert-butoxycarbonyl amino-2-methylpropanoic acid, boc-2-aminoisobutyric acid, n-boc-2-aminoisobutyric acid, n-boc-alpha-methylalanine, n-boc-2-amino-2-methylpropanoic acid, 2-tert-butoxy carbonyl amino-2-methylpropanoic acid, 2-tert-butoxycarbonylamino isobutyric acid
PubChem CID 2733845
IUPAC Name 2-methyl-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
SMILES CC(C)(C)OC(=O)NC(C)(C)C(=O)O

Specifications

Melting Point 118°C to 121°C
Beilstein 1953772
Quantity 1 g
Formula Weight 203.24
Percent Purity ≥98%
Chemical Name or Material N-Boc-2-aminoisobutyric acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.