missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-1H-pyrazole-1-carboxamidine, 98+%

Product Code. 15490698
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15490698 25 g 25g
15470698 1 g 1g
15480698 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15490698 Supplier Thermo Scientific Alfa Aesar Supplier No. H54394.14
 Restricted Item
Estimated Shipment: 04-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 152120-61-1 | C9H14N4O2 | 210.237 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 152120-61-1
Molecular Formula C9H14N4O2
Molecular Weight (g/mol) 210.237
MDL Number MFCD00216663
InChI Key IGSFMHYSWZUENI-UHFFFAOYSA-N
Synonym n-boc-1-guanylpyrazole, n-boc-1h-pyrazole-1-carboxamidine, n-boc-pyrazole-1-carboxamidine, n-tert-butoxycarbonyl-1h-pyrazole-1-carboxamidine, tert-butyl 1e-amino 1h-pyrazol-1-yl methylene carbamate, n-tert-butoxycarbonyl-1 h-pyrazole-1-carboxamidine, h-pyrazole-1-carboxamidine hcl, 1h-pyrazole-n-t-butyloxycarbonyl-1-carboxamidine, tert-butoxy-n-iminopyrazolylmethyl carboxamide, tert-butyl imino 1h-pyrazol-1-yl methylcarbamate
PubChem CID 9605068
IUPAC Name tert-butyl (NE)-N-[amino(pyrazol-1-yl)methylidene]carbamate
SMILES CC(C)(C)OC(=O)N=C(N)N1C=CC=N1

Specifications

Melting Point 97°C to 101°C
Quantity 25 g
Beilstein 6141956
Formula Weight 210.23
Percent Purity ≥98%
Chemical Name or Material N-Boc-1H-pyrazole-1-carboxamidine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.