missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-1-benzyloxymethyl-L-histidine, 97%, Thermo Scientific™

Product Code. 15499158
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
This item is not returnable. View return policy

Product Code. 15499158

Brand: Thermo Scientific Alfa Aesar H62663.MD

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 83468-83-1
Molecular Formula C19H25N3O5
Molecular Weight (g/mol) 375.43
MDL Number MFCD00038627
InChI Key IEAOAWZLUGOPJX-UHFFFAOYNA-N
Synonym boc-l-his bom-oh, n-boc-n'-benzyloxymethyl-l-histidine, s-3-1-benzyloxy methyl-1h-imidazol-4-yl-2-tert-butoxycarbonyl amino propanoic acid, bochis bom oh, pubchem18939, n alpha-tert-butyloxycarbonyl-n pi-benzyloxyhistidine, l-histidine, n-1,1-dimethylethoxy carbonyl-1-phenylmethoxy methyl, 2s-3-1-benzyloxymethyl imidazol-4-yl-2-tert-butoxycarbonylamino propanoic acid, 2s-3-1-benzyloxy methyl imidazol-4-yl-2-tert-butoxycarbonyl amino propanoic acid
PubChem CID 11245813
IUPAC Name (2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-3-[1-(phenylmethoxymethyl)imidazol-4-yl]propanoic acid
SMILES CC(C)(C)OC(=O)NC(CC1=CN(COCC2=CC=CC=C2)C=N1)C(O)=O

Specifications

Color White
Quantity 250 mg
Formula Weight 375.42
Percent Purity 97%
Physical Form Powder
Chemical Name or Material N-Boc-1-benzyloxymethyl-L-histidine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.