missing translation for 'onlineSavingsMsg'
Learn More

N-BOC-1-Aminocyclobutanecarboxylic acid, 97%, Thermo Scientific Chemicals

Product Code. 11925501 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11925501 5 g 5g
11935501 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11925501 Supplier Thermo Scientific Acros Supplier No. 440440050

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 120728-10-1
Molecular Formula C10H16NO4
Molecular Weight (g/mol) 214.24
MDL Number MFCD02682623
InChI Key ROVVUKFHORPDSM-UHFFFAOYSA-M
Synonym n-boc-1-aminocyclobutanecarboxylic acid, 1-tert-butoxycarbonyl amino cyclobutanecarboxylic acid, 1-boc-amino cyclobutanecarboxylic acid, boc-cyclovaline, boc-acbc-oh, 1-n-boc-amino-cyclobutane carboxylic acid, 1-tert-butoxycarbonylamino cyclobutanecarboxylic acid, boc-1-aminocyclobutane-1-carboxylic acid, n-boc-1-aminocyclobutane carboxylic acid, boc-1-aminocyclobutanecarboxylic acid
PubChem CID 1512646
IUPAC Name 1-[(2-methylpropan-2-yl)oxycarbonylamino]cyclobutane-1-carboxylic acid
SMILES CC(C)(C)OC(=O)NC1(CCC1)C([O-])=O

Specifications

Melting Point 129°C to 133°C
CAS Min % 96.0
CAS Max % 100.0
Color White to Brown
Infrared Spectrum Authentic
Assay Percent Range 97%
Quantity 5 g
Formula Weight 215.25
Percent Purity 97%
Physical Form Powder
Chemical Name or Material N-BOC-1-Aminocyclobutanecarboxylic acid
Show More Show Less
compliance-icons
Product Identifier
  • N-BOC-1-Aminocyclobutanecarboxylic acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P264-Wash face, hands and any thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.