missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-1,6-diaminohexane hydrochloride, 98%

Product Code. 11358836
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11358836 1 g 1g
11368836 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11358836 Supplier Thermo Scientific Alfa Aesar Supplier No. L19487.03
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 65915-94-8 | C11H25ClN2O2 | 252.78 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 65915-94-8
Molecular Formula C11H25ClN2O2
Molecular Weight (g/mol) 252.78
MDL Number MFCD00039072
InChI Key JSBWQIZQJOQPFN-UHFFFAOYSA-N
Synonym n-boc-1,6-hexanediamine hydrochloride, tert-butyl 6-aminohexyl carbamate hydrochloride, n-boc-1,6-diamino-hexane hydrochloride, tert-butyl n-6-aminohexyl carbamate hydrochloride, n-6-aminohexyl tert-butoxy carboxamide, chloride, tert-butyl 6-aminohexyl carbamate monohydrochloride, tert-butyl 6-aminohexyl carbamate hcl, acmc-1b5cm, n-1-boc-1,6-diaminohexane . hcl, n-boc-1,6-hexanediamine hcl
PubChem CID 3017638
IUPAC Name tert-butyl N-(6-aminohexyl)carbamate;hydrochloride
SMILES [H+].[Cl-].CC(C)(C)OC(=O)NCCCCCCN

Specifications

Melting Point ∽164°C (decomposition)
Beilstein 3631740
Quantity 1 g
Formula Weight 252.79
Sensitivity Hygroscopic
Percent Purity 98%
Chemical Name or Material N-Boc-1,6-diaminohexane hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.