missing translation for 'onlineSavingsMsg'
Learn More

N-Bis(2-hydroxypropyl)nitrosamine, MedChemExpress

Product Code. 16494558
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
25 mg
50 mg
100 mg
200 mg
Unit Size:
100mg
10mg
200mg
25mg
50mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16494558 10 mg 10mg
16473126 25 mg 25mg
16471016 200 mg 200mg
16425416 100 mg 100mg
16420867 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16494558 Supplier Medchem Express Supplier No. HY112085
 Restricted Item
Estimated Shipment: 29-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress N-Bis(2-hydroxypropyl)nitrosamine is an agent with carcinogenic activity.

N-Bis(2-hydroxypropyl)nitrosamine is an agent with carcinogenic activity.

Specifications

Chemical Name or Material N-Bis(2-hydroxypropyl)nitrosamine
Physical Form Oil
CAS 53609-64-6
Color Light Yellow
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C6H14N2O3
For Use With (Application) Cancer-programmed cell death
Synonym DHPN Di(2-hydroxypropyl)nitrosamine Diisopropanolnitrosamine
Solubility Information DMSO : ≥ 125 mg/mL (770.70 mM)
SMILES O=NN(CC(O)C)CC(O)C
Molecular Weight (g/mol) 162.19
Formula Weight 162.19
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.