missing translation for 'onlineSavingsMsg'
Learn More

N-[Beta-Hydroxy-Beta-(4-hydroxyphenyl)ethyl] Ractopamine(Mixture of Diastereomers), TRC

Product Code. 30392666
Change view
Click to view available options
Quantity:
1 mg
2.5 mg
25 mg
Unit Size:
2.5mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30392666 2.5 mg 2.5mg
30389186 25 mg 25mg
30397421 1 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30392666 Supplier Toronto Research Chemicals Supplier No. TRCH8299902.5MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1797121-82-4
Molecular Formula C26H31NO5
Molecular Weight (g/mol) 437.53
IUPAC Name 4-[3-[bis[2-hydroxy-2-(4-hydroxyphenyl)ethyl]amino]butyl]phenol
SMILES CC(CCc1ccc(O)cc1)N(CC(O)c2ccc(O)cc2)CC(O)c3ccc(O)cc3

Specifications

Quantity 2.5 mg
InChI Formula InChI=1S/C26H31NO5/c1-18(2-3-19-4-10-22(28)11-5-19)27(16-25(31)20-6-12-23(29)13-7-20)17-26(32)21-8-14-24(30)15-9-21/h4-15,18,25-26,28-32H,2-3,16-17H2,1H3
Formula Weight 437.2202
Chemical Name or Material N-[beta-Hydroxy-beta-(4-hydroxyphenyl)ethyl] Ractopamine(Mixture of Diastereomers)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.