missing translation for 'onlineSavingsMsg'
Learn More

N-(Beta-Chloroethyl)piperidine Hydrochloride, TRC

Product Code. 30362927
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30362927 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30362927 Supplier Toronto Research Chemicals Supplier No. TRCC36674525G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-(beta-Chloroethyl)piperidine Hydrochloride
CAS 2008-75-5
Molecular Formula C7H14Cl2N
Quantity 25 g
Synonym 1-(2-Chloroethyl)piperidine Hydrochloride, 1-(2-Chloroethyl)piperidine Monohydrochloride, 1-(2-Chloroethyl)piperidinium Chloride, 1-Chloro-2-(1-piperidinyl)ethane Monohydrochloride, 1-Chloro-2-piperidinoethane Hydrochloride, 2-(1-Piperidinyl)-1-chloroethane Hydrochloride, 2-(Piperidin-1-yl)ethyl Chloride Hydrochloride, 2-Piperidino-1-chloroethane Hydrochloride, 2-Piperidinoethyl Chloride Hydrochloride, N-(2-Chloroethyl)piperidine Hydrochloride
InChI Formula InChI=1S/C7H14ClN.ClH/c8-4-7-9-5-2-1-3-6-9;/h1-7H2;1H
SMILES Cl.ClCCN1CCCCC1
IUPAC Name 1-(2-chloroethyl)piperidine;hydrochloride
Molecular Weight (g/mol) 183.1
Formula Weight 183.0582
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.