missing translation for 'onlineSavingsMsg'
Learn More

N-Benzyltropinone, 97%, Thermo Scientific™

Product Code. 10081524
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Unit Size:
10g
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10081524 1 g 1g
10159152 10 g 10g
10595544 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10081524 Supplier Thermo Scientific Maybridge Supplier No. MO07738DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 28957-72-4
Molecular Formula C14H17NO
Molecular Weight (g/mol) 215.30
MDL Number MFCD00151640
InChI Key RSUHKGOVXMXCND-UHFFFAOYNA-N
Synonym 8-benzyl-8-azabicyclo 3.2.1 octan-3-one, n-benzylnortropinone, n-benzyltropinone, 8-benzyl-8-aza-bicyclo 3.2.1 octan-3-one, 8-phenylmethyl-8-azabicyclo 3.2.1 octan-3-one, 8-azabicyclo 3.2.1 octan-3-one, 8-phenylmethyl, n-benzyl tropinone, 8-benzyl-8-azabicyclo 3.2.1 octan-3-on, n-benzyl-8-azabicyclo 3.2.1 octan-3-one
PubChem CID 119846
IUPAC Name 8-benzyl-8-azabicyclo[3.2.1]octan-3-one
SMILES O=C1CC2CCC(C1)N2CC1=CC=CC=C1

Specifications

Assay Percent Range 97%
Quantity 1 g
Formula Weight 215.29
Percent Purity 97%
Chemical Name or Material 8-Benzyl-8-azabicyclo[3.2.1]octan-3-one
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.