missing translation for 'onlineSavingsMsg'
Learn More

N-Benzyltrifluoromethanesulfonamide, 97%

Product Code. 11309345
Click to view available options
Quantity:
25 g
Unit Size:
25g
This item is not returnable. View return policy

Product Code. 11309345

Brand: Thermo Scientific Alfa Aesar L16705.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 36457-58-6 | C8H8F3NO2S | 239.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 36457-58-6
Molecular Formula C8H8F3NO2S
Molecular Weight (g/mol) 239.21
MDL Number MFCD00042074
InChI Key IJHVVEQTOXFGCL-UHFFFAOYSA-N
PubChem CID 118985
IUPAC Name N-benzyl-1,1,1-trifluoromethanesulfonamide
SMILES FC(F)(F)S(=O)(=O)NCC1=CC=CC=C1

Specifications

Melting Point 44°C to 46°C
Linear Formula C6H5CH2NHSO2CF3
Quantity 25 g
Beilstein 2119124
Synonym n-benzyltrifluoromethanesulfonamide, n-benzyltriflamide, n-benzyl trifluoromethanesulfonamide, n-benzyl-1,1,1-trifluoromethanesulphonamide, benzyl trifluoromethyl sulfonyl amine, trifluoromethanesulfonamidenbenzyl, acmc-1ahjf, 1,1,1-trifluoro-n-phenylmethyl methanesulfonamide, methanesulfonamide,1,1,1-trifluoro-n-phenylmethyl, methanesulfonamide, 1,1,1-trifluoro-n-phenylmethyl
Formula Weight 239.22
Percent Purity 97%
Chemical Name or Material N-Benzyltrifluoromethanesulfonamide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.