missing translation for 'onlineSavingsMsg'
Learn More

N-Benzyloxycarbonylhydroxylamine, TRC

Product Code. 30331551
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30331551 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30331551 Supplier Toronto Research Chemicals Supplier No. TRCB2859155G
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-Benzyloxycarbonylhydroxylamine
CAS 3426-71-9
Molecular Formula C8 H9 N O3
Quantity 5 g
Synonym Carbamic acid, N-hydroxy-, phenylmethyl ester, Carbamic acid, hydroxy-, benzyl ester (6CI,7CI,8CI), Carbamic acid, hydroxy-, phenylmethyl ester (9CI), Benzyl N-hydroxycarbamate, Benzyl hydroxycarbamate, N-Benzyloxycarbonylhydroxylamine, N-Cbz-hydroxylamine, N-Hydroxycarbamic acid benzyl ester, NSC 528506, N-Hydroxycarbamic acid phenylmethyl ester
InChI Formula InChI=1S/C8H9NO3/c10-8(9-11)12-6-7-4-2-1-3-5-7/h1-5,11H,6H2,(H,9,10)
SMILES ONC(=O)OCc1ccccc1
IUPAC Name benzyl N-hydroxycarbamate
Molecular Weight (g/mol) 167.16
Formula Weight 167.0582
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.