missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Benzyloxycarbonylglycine methyl ester, 98%, Thermo Scientific Chemicals
£30.80 - £389.00
Chemical Identifiers
CAS | 1212-53-9 |
---|---|
Molecular Formula | C11H13NO4 |
Molecular Weight (g/mol) | 223.23 |
MDL Number | MFCD00043631 |
InChI Key | DZYBBBYFLOPVOL-UHFFFAOYSA-N |
Synonym | z-gly-ome, z-glycine methyl ester, methyl 2-benzyloxy carbonyl amino acetate, carbobenzyloxyglycine methyl ester, n-cbz-glycine methyl ester, methyl n-benzyloxycarbonylglycinate, cbz-glycine methyl ester, carbobenzoxyglycine methyl ester, methyl 2-benzyloxycarbonylamino acetate, glycine, n-phenylmethoxy carbonyl-, methyl ester |
PubChem CID | 71033 |
SMILES | COC(=O)CNC(=O)OCC1=CC=CC=C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15479308
|
Thermo Scientific Alfa Aesar
H62858.06 |
5 g |
£30.80
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15489308
|
Thermo Scientific Alfa Aesar
H62858.14 |
25 g |
£112.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15499308
|
Thermo Scientific Alfa Aesar
H62858.22 |
100 g | |||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1212-53-9 | |
223.23 | |
DZYBBBYFLOPVOL-UHFFFAOYSA-N | |
71033 |
C11H13NO4 | |
MFCD00043631 | |
z-gly-ome, z-glycine methyl ester, methyl 2-benzyloxy carbonyl amino acetate, carbobenzyloxyglycine methyl ester, n-cbz-glycine methyl ester, methyl n-benzyloxycarbonylglycinate, cbz-glycine methyl ester, carbobenzoxyglycine methyl ester, methyl 2-benzyloxycarbonylamino acetate, glycine, n-phenylmethoxy carbonyl-, methyl ester | |
COC(=O)CNC(=O)OCC1=CC=CC=C1 |
Specifications
1212-53-9 | |
147°C to 151°C | |
C6H5CH2OCONHCH2CO2CH3 | |
z-gly-ome, z-glycine methyl ester, methyl 2-benzyloxy carbonyl amino acetate, carbobenzyloxyglycine methyl ester, n-cbz-glycine methyl ester, methyl n-benzyloxycarbonylglycinate, cbz-glycine methyl ester, carbobenzoxyglycine methyl ester, methyl 2-benzyloxycarbonylamino acetate, glycine, n-phenylmethoxy carbonyl-, methyl ester | |
COC(=O)CNC(=O)OCC1=CC=CC=C1 | |
223.23 | |
223.23 | |
N-Benzyloxycarbonylglycine methyl ester |
22°C to 26°C | |
C11H13NO4 | |
MFCD00043631 | |
DZYBBBYFLOPVOL-UHFFFAOYSA-N | |
methyl 2-(phenylmethoxycarbonylamino)acetate | |
71033 | |
98% |
Safety and Handling
missing translation for 'einecsNumber' : 214-922-8
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only