missing translation for 'onlineSavingsMsg'
Learn More

N-Benzyloxycarbonylglycine, 98+%

Product Code. 11458466
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11458466 25 g 25g
11468466 100 g 100g
11478466 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11458466 Supplier Thermo Scientific Alfa Aesar Supplier No. A15928.14
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 1138-80-3 | C10H11NO4 | 209.201 g/mol

N-Carbobenzoxyglycine is used in the dipeptide synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Carbobenzoxyglycine is used in the dipeptide synthesis.

Solubility
Soluble in methanol. Insoluble in water.

Notes
Store in cool dry place. Keep away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1138-80-3
Molecular Formula C10H11NO4
Molecular Weight (g/mol) 209.201
MDL Number MFCD00002691
InChI Key CJUMAFVKTCBCJK-UHFFFAOYSA-N
Synonym n-carbobenzyloxyglycine, n-cbz-glycine, z-gly-oh, carbobenzyloxyglycine, carbobenzoxyglycine, n-benzyloxycarbonylglycine, n-carbobenzoxyglycine, cbz-gly-oh, cbz gly, benzyloxycarbonylglycine
PubChem CID 14349
ChEBI CHEBI:16532
IUPAC Name 2-(phenylmethoxycarbonylamino)acetic acid
SMILES C1=CC=C(C=C1)COC(=O)NCC(=O)O

Specifications

Melting Point 119°C to 122°C
Linear Formula C6H5CH2OCONHCH2CO2H
Beilstein 526877
Solubility Information Soluble in methanol. Insoluble in water.
Quantity 25 g
Formula Weight 209.2
Percent Purity ≥98%
Chemical Name or Material N-Benzyloxycarbonylglycine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.