missing translation for 'onlineSavingsMsg'
Learn More

N-(Benzyloxycarbonyl)serine Benzyl Ester, TRC

Product Code. 30340521
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30340521 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30340521 Supplier Toronto Research Chemicals Supplier No. TRCB28677010G
 Restricted Item
Estimated Shipment: 24-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-(Benzyloxycarbonyl)serine Benzyl Ester
CAS 21209-51-8
Molecular Formula C18H19NO5
Quantity 10 g
Synonym (Benzyloxycarbonyl)serine Benzyl Ester, Benzyl (S)-(-)-2-Benzyloxycarbonylamino-3-hydroxypropionate, N-(Benzyloxycarbonyl)-L-serine Benzyl Ester, N-Carboxyserine DIbenzyl Ester, N-[(Phenylmethoxy)carbonyl]-L-serine Phenylmethyl Ester
InChI Formula InChI=1S/C18H19NO5/c20-11-16(17(21)23-12-14-7-3-1-4-8-14)19-18(22)24-13-15-9-5-2-6-10-15/h1-10,16,20H,11-13H2,(H,19,22)/t16-/m0/s1
SMILES OC[C@H](NC(=O)OCc1ccccc1)C(=O)OCc2ccccc2
IUPAC Name benzyl (2S)-3-hydroxy-2-(phenylmethoxycarbonylamino)propanoate
Molecular Weight (g/mol) 329.35
Formula Weight 329.1263
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.