missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Benzyloxycarbonyl-O-benzyl-L-tyrosine, 95%, Thermo Scientific™
£15.90 - £54.10
Chemical Identifiers
CAS | 16677-29-5 |
---|---|
Molecular Formula | C24H23NO5 |
Molecular Weight (g/mol) | 405.45 |
MDL Number | MFCD00037819 |
InChI Key | IPAODWFPTVIUSZ-ANBDAQEENA-N |
Synonym | z-tyr bzl-oh, z-o-benzyl-l-tyrosine, cbz-tyr bzl-oh, cbz-o-benzyl-l-tyrosine, s-2-benzyloxy carbonyl amino-3-4-benzyloxy phenyl propanoic acid, l-tyrosine, n-phenylmethoxy carbonyl-o-phenylmethyl, 2s-2-benzyloxy carbonyl amino-3-4-benzyloxy phenyl propanoic acid, z-tyr bzl, cbz-o-benzyl-l-tyr, pubchem19061 |
PubChem CID | 47600 |
IUPAC Name | (2S)-2-(phenylmethoxycarbonylamino)-3-(4-phenylmethoxyphenyl)propanoic acid |
SMILES | OC(=O)[C@H](CC1=CC=C(OCC2=CC=CC=C2)C=C1)NC(=O)OCC1=CC=CC=C1 |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
16677-29-5 | |
405.45 | |
IPAODWFPTVIUSZ-ANBDAQEENA-N | |
47600 | |
OC(=O)[C@H](CC1=CC=C(OCC2=CC=CC=C2)C=C1)NC(=O)OCC1=CC=CC=C1 |
C24H23NO5 | |
MFCD00037819 | |
z-tyr bzl-oh, z-o-benzyl-l-tyrosine, cbz-tyr bzl-oh, cbz-o-benzyl-l-tyrosine, s-2-benzyloxy carbonyl amino-3-4-benzyloxy phenyl propanoic acid, l-tyrosine, n-phenylmethoxy carbonyl-o-phenylmethyl, 2s-2-benzyloxy carbonyl amino-3-4-benzyloxy phenyl propanoic acid, z-tyr bzl, cbz-o-benzyl-l-tyr, pubchem19061 | |
(2S)-2-(phenylmethoxycarbonylamino)-3-(4-phenylmethoxyphenyl)propanoic acid |
Specifications
16677-29-5 | |
MFCD00037819 | |
z-tyr bzl-oh, z-o-benzyl-l-tyrosine, cbz-tyr bzl-oh, cbz-o-benzyl-l-tyrosine, s-2-benzyloxy carbonyl amino-3-4-benzyloxy phenyl propanoic acid, l-tyrosine, n-phenylmethoxy carbonyl-o-phenylmethyl, 2s-2-benzyloxy carbonyl amino-3-4-benzyloxy phenyl propanoic acid, z-tyr bzl, cbz-o-benzyl-l-tyr, pubchem19061 | |
OC(=O)[C@H](CC1=CC=C(OCC2=CC=CC=C2)C=C1)NC(=O)OCC1=CC=CC=C1 | |
405.45 | |
405.44 | |
Solid |
C24H23NO5 | |
3178475 | |
IPAODWFPTVIUSZ-ANBDAQEENA-N | |
(2S)-2-(phenylmethoxycarbonylamino)-3-(4-phenylmethoxyphenyl)propanoic acid | |
47600 | |
95% | |
N-Benzyloxycarbonyl-O-benzyl-L-tyrosine |
Safety and Handling
missing translation for 'tsca' : No
missing translation for 'storageNote' : 2° to 8°C
Recommended Storage : Keep cold