missing translation for 'onlineSavingsMsg'
Learn More

N-Benzyloxycarbonyl-L-tyrosine methyl ester, 98%

Product Code. 15418136
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 15418136

Brand: Thermo Scientific Alfa Aesar H63113.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13512-31-7 | C18H19NO5 | 329.35 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13512-31-7
Molecular Formula C18H19NO5
Molecular Weight (g/mol) 329.35
MDL Number MFCD00038784
InChI Key SLLMDHBKALJDBW-XISACWJONA-N
Synonym z-tyr-ome, z-l-tyrosine methyl ester, n-cbz-l-tyrosine methyl ester, s-methyl 2-benzyloxy carbonyl amino-3-4-hydroxyphenyl propanoate, cbz-tyr-ome, z-l-tyr-ome, z-tyr-och3, cbz-tyrosine methyl ester, n-cbz-l-tyrosinemethylester, n-benzyloxycarbonyltyrosine methyl ester
PubChem CID 7010613
SMILES COC(=O)[C@H](CC1=CC=C(O)C=C1)NC(=O)OCC1=CC=CC=C1

Specifications

Melting Point 93°C to 97°C (decomposition)
IUPAC Name methyl 2-{[(benzyloxy)carbonyl]amino}-3-(4-hydroxyphenyl)propanoate
Quantity 100 g
Formula Weight 329.35
Percent Purity 98%
Physical Form Solid
Chemical Name or Material N-Benzyloxycarbonyl-L-tyrosine methyl ester

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.