missing translation for 'onlineSavingsMsg'
Learn More

N-Benzyloxycarbonyl-L-threonine, 99%

Product Code. 11305109
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11305109 5 g 5g
11315109 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11305109 Supplier Thermo Scientific Alfa Aesar Supplier No. A17565.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 19728-63-3 | C12H15NO5 | 253.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19728-63-3
Molecular Formula C12H15NO5
Molecular Weight (g/mol) 253.25
MDL Number MFCD00065948
InChI Key IPJUIRDNBFZGQN-SCZZXKLOSA-N
Synonym z-thr-oh, n-cbz-l-threonine, z-l-threonine, z-l-thr-oh, n-benzyloxycarbonyl-l-threonine, cbz-l-threonine, n-carbobenzyloxy-l-threonine, l-threonine, n-phenylmethoxy carbonyl, benzyloxycarbonyl-l-threonine, n-benzyloxy carbonyl-l-threonine
PubChem CID 88217
IUPAC Name (2S,3R)-3-hydroxy-2-(phenylmethoxycarbonylamino)butanoic acid
SMILES C[C@@H](O)[C@H](NC(=O)OCC1=CC=CC=C1)C(O)=O

Specifications

Melting Point 103°C to 105°C
Quantity 5 g
Beilstein 2335409
Optical Rotation −4.5° (c=4 in Acetic Acid)
Formula Weight 253.26
Percent Purity 99%
Chemical Name or Material N-Benzyloxycarbonyl-L-threonine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.